3-Fluoro-6-methoxyquinoline ≥97%

Supplier: Matrix Scientific

087254-500MG
102853-088EA 1086.15 USD
102853-088
3-Fluoro-6-methoxyquinoline ≥97%
3-Fluoro-6-methoxyquinoline

Hazard: Irritant

Formula: C₁₀H₈FNO
MDL Number: MFCD12400834
CAS Number: 426842-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR