4-(Chloromethyl)-1-fluoro-2-iodobenzene ≥95%

Supplier: Matrix Scientific

087235-1G 087235-500MG
102853-008EA 1504.62 USD
102853-008 102853-006
4-(Chloromethyl)-1-fluoro-2-iodobenzene ≥95%
4-(Chloromethyl)-1-fluoro-2-iodobenzene

Hazard: Irritant

Formula: C₇H₅ClFI
MDL Number: MFCD18396969
CAS Number: 1261837-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR