6-Benzyl-2,6-diazaspiro[4.5]decane ≥97%

Supplier: Matrix Scientific

087215-500MG
102852-930EA 2396.92 USD
102852-930
6-Benzyl-2,6-diazaspiro[4.5]decane ≥97%
6-Benzyl-2,6-diazaspiro[4.5]decane

Hazard: Irritant

Formula: C₁₅H₂₂N₂
MDL Number: MFCD12406552
CAS Number: 1158750-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR