2-(Benzenesulfonyl)-N,N-diethylacetamide ≥97%
Supplier: Matrix Scientific
087214-500MG
102852-928EA
403.08
USD
102852-928
2-(Benzenesulfonyl)-N,N-diethylacetamide ≥97%
2-(Benzenesulfonyl)-N,N-diethylacetamide
Hazard: Irritant
Formula:
C₁₂H₁₇NO₃S MW: 255.34 g/mol |
MDL Number:
MFCD04274826 CAS Number: 163019-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...