5-Acetyl-3-iodo-6-methyl-1,2-dihydropyridin-2-one ≥95%
Supplier: Matrix Scientific
087209-500MG
087209-1G
102852-908EA
803.08
USD
102852-908
102852-910
5-Acetyl-3-iodo-6-methyl-1,2-dihydropyridin-2-one ≥95%
5-Acetyl-3-iodo-6-methyl-1,2-dihydropyridin-2-one
Hazard: Irritant
Formula:
C₈H₈INO₂ Melting Pt: 223…226 °C |
MDL Number:
MFCD22573522 CAS Number: 1407532-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...