(3-Methyl-4-nitrophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3-Methyl-4-nitrophenyl)acetic acid

087181-5G 087181-1G
102852-874EA 98.46 USD
102852-874 102852-876
(3-Methyl-4-nitrophenyl)acetic acid ≥95%
(3-Methyl-4-nitrophenyl)acetic acid

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD11036394
CAS Number: 143665-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR