6-Chloro-8-fluoroquinoline ≥95%

Supplier: Matrix Scientific

087175-1G
102852-864EA 173.85 USD
102852-864
6-Chloro-8-fluoroquinoline ≥95%
6-Chloro-8-fluoroquinoline

Hazard: Irritant

Formula: C₉H₅ClFN
Melting Pt: 79…81 °C
MDL Number: MFCD16658595
CAS Number: 52200-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR