2-Phenylimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

087171-1G
102852-856EA 169.23 USD
102852-856
2-Phenylimidazo[1,2-a]pyridine ≥95%
2-Phenylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD00219254
CAS Number: 4105-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR