3-(1,3-Dioxan-2-yl)-4'-trifluoromethylpropiophenone ≥97%

Supplier: Matrix Scientific

087153-2G 087153-5G 087153-1G
102852-814EA 712.31 USD
102852-814 102852-816 102852-818
3-(1,3-Dioxan-2-yl)-4'-trifluoromethylpropiophenone ≥97%
3-(1,3-Dioxan-2-yl)-4'-trifluoromethylpropiophenone

Hazard: Irritant

Formula: C₁₄H₁₅F₃O₃
Melting Pt: 46…47 °C
MDL Number: MFCD02261808
CAS Number: 898786-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR