1-Bromo-5-(1,3-dioxolan-2-yl)thiophene ≥97%

Supplier: Matrix Scientific

087147-2G 087147-1G 087147-5G
102852-780EA 752.31 USD
102852-780 102852-782 102852-778
1-Bromo-5-(1,3-dioxolan-2-yl)thiophene ≥97%
1-Bromo-5-(1,3-dioxolan-2-yl)thiophene

Hazard: Irritant

Formula: C₇H₇BrO₂S
MDL Number: MFCD06209038
CAS Number: 52157-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR