4-Phenylbutyraldehyde ≥97%
Supplier: Matrix Scientific
087145-2G
087145-5G
087145-1G
102852-770EA
1760
USD
102852-770
102852-766
102852-768
4-Phenylbutyraldehyde ≥97%
4-Phenylbutyraldehyde
Hazard: Irritant/Refrigerate
Formula:
C₁₀H₁₂O |
MDL Number:
MFCD00800383 CAS Number: 18328-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...