3-(3-Chloro-5-fluorophenyl)propionaldehyde ≥97%
Supplier: Matrix Scientific
087144-2G
087144-1G
087144-5G
102852-760EA
490.77
USD
102852-760
102852-762
102852-764
3-(3-Chloro-5-fluorophenyl)propionaldehyde ≥97%
3-(3-Chloro-5-fluorophenyl)propionaldehyde
Hazard: Irritant
Formula:
C₉H₈ClFO |
MDL Number:
MFCD08741517 CAS Number: 951890-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...