3-(Dimethylamino)benzaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(N,N-Dimethylamino)benzaldehyde

087143-2G 087143-5G 087143-1G
102852-754EA 769.23 USD
102852-754 102852-756 102852-758
3-(Dimethylamino)benzaldehyde ≥97%
3-(Dimethylamino)benzaldehyde

Hazard: Irritant/Stored under Argon

Formula: C₉H₁₁NO
MW: 149.1897 g/mol
MDL Number: MFCD05148649
CAS Number: 619-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR