3-Cyclohexylpropionaldehyde ≥97%

Supplier: Matrix Scientific

087141-5G 087141-2G 087141-1G
102852-742EA 769.23 USD
102852-742 102852-744 102852-746
3-Cyclohexylpropionaldehyde ≥97%
3-Cyclohexylpropionaldehyde

Hazard: Irritant

Formula: C₉H₁₆O
MDL Number: MFCD00012280
CAS Number: 4361-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR