3-(2-Methylphenyl)propionaldehyde ≥95%

Supplier: Matrix Scientific

087139-1G 087139-5G 087139-2G
102852-730EA 769.23 USD
102852-730 102852-732 102852-734
3-(2-Methylphenyl)propionaldehyde ≥95%
3-(2-Methylphenyl)propionaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O
MDL Number: MFCD07772906
CAS Number: 19564-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR