1-Bromo-2-(1,3-dioxolan-2-ylmethyl)benzene ≥97%
Supplier: Matrix Scientific
087126-5G
087126-1G
087126-2G
102852-652EA
490.77
USD
102852-652
102852-654
102852-656
1-Bromo-2-(1,3-dioxolan-2-ylmethyl)benzene ≥97%
1-Bromo-2-(1,3-dioxolan-2-ylmethyl)benzene
Hazard: Irritant
Formula:
C₁₀H₁₁BrO₂ |
MDL Number:
MFCD04117643 CAS Number: 96557-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...