Ethyl-3-oxocyclopentane-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-oxocyclopentanecarboxylate

087098-1G 087098-5G 087098-10G
102852-484EA 315.38 USD
102852-484 102852-486 102852-488
Ethyl-3-oxocyclopentane-1-carboxylate ≥97%
Ethyl-3-oxocyclopentane-1-carboxylate

Hazard: Irritant

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
MDL Number: MFCD01320352
CAS Number: 5400-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR