Ethyl-7-oxodecanoate ≥97%

Supplier: Matrix Scientific

087096-2G 087096-1G 087096-5G
102852-472EA 592.31 USD
102852-472 102852-474 102852-476
Ethyl-7-oxodecanoate ≥97%
Ethyl-7-oxodecanoate

Hazard: Irritant

Formula: C₁₂H₂₂O₃
MDL Number: MFCD01320350
CAS Number: 73276-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR