Ethyl-5-oxodecanoate ≥97%

Supplier: Matrix Scientific

087092-2G 087092-5G 087092-1G
102852-450EA 944.62 USD
102852-450 102852-452 102852-448
Ethyl-5-oxodecanoate ≥97%
Ethyl-5-oxodecanoate

Hazard: Irritant

Formula: C₁₂H₂₂O₃
MDL Number: MFCD01320344
CAS Number: 93919-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR