Ethyl-5-cyclopentyl-5-oxovalerate ≥97%
Supplier: Matrix Scientific
087072-2G
087072-5G
087072-1G
102852-330EA
944.62
USD
102852-330
102852-332
102852-328
Ethyl-5-cyclopentyl-5-oxovalerate ≥97%
Ethyl-5-cyclopentyl-5-oxovalerate
Hazard: Irritant
Formula:
C₁₂H₂₀O₃ |
MDL Number:
MFCD01320323 CAS Number: 24071-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...