Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(4-Methylphenyl)-4-oxobutanoic acid ethyl ester
, 4-Oxo-4-(p-tolyl)butyric acid ethyl ester
, Ethyl 4-Oxo-4-(p-tolyl)butyrate
, Ethyl 4-(4-Methylphenyl)-4-oxobutanoate
087063-2G
087063-5G
087063-1G
102852-274EA
886.15
USD
102852-274
102852-276
102852-278
Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥97%
Ethyl-4-(4-methylphenyl)-4-oxobutyrate
Hazard: Irritant
Formula:
C₁₃H₁₆O₃ MW: 220.27 g/mol |
MDL Number:
MFCD00266564 CAS Number: 6942-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...