4-(3-Ethoxyphenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087050-1G 087050-2G 087050-5G
102852-196EA 490.77 USD
102852-196 102852-198 102852-200
4-(3-Ethoxyphenyl)-4-oxobutyric acid ≥97%
4-(3-Ethoxyphenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MDL Number: MFCD09801791
CAS Number: 905592-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR