4-Oxo-4-(2-trifluoromethylphenyl)butyric acid ≥97%
Supplier: Matrix Scientific
087037-2G
087037-1G
087037-5G
102852-120EA
944.62
USD
102852-120
102852-122
102852-118
4-Oxo-4-(2-trifluoromethylphenyl)butyric acid ≥97%
4-Oxo-4-(2-trifluoromethylphenyl)butyric acid
Hazard: Irritant
Formula:
C₁₁H₉F₃O₃ |
MDL Number:
MFCD00033285 CAS Number: 200725-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...