3-(1,3-Dioxolan-2-ylmethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087032-1G 087032-2G 087032-5G
102852-088EA 592.31 USD
102852-088 102852-090 102852-092
3-(1,3-Dioxolan-2-ylmethyl)benzoic acid ≥97%
3-(1,3-Dioxolan-2-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₄
MDL Number: MFCD06200652
CAS Number: 898767-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR