2-(2-Oxopropyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087028-1G 087028-2G 087028-5G
102852-064EA 592.31 USD
102852-064 102852-066 102852-068
2-(2-Oxopropyl)benzoic acid ≥97%
2-(2-Oxopropyl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MDL Number: MFCD02260862
CAS Number: 2852-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR