3-Propionylbenzoic acid ≥97%

Supplier: Matrix Scientific

087026-1G 087026-5G 087026-2G
102852-052EA 490.77 USD
102852-052 102852-054 102852-056
3-Propionylbenzoic acid ≥97%
3-Propionylbenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MDL Number: MFCD02260860
CAS Number: 74380-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR