5-Oxo-5-(2-pyridyl)valeric acid ≥97%

Supplier: Matrix Scientific

087018-1G 087018-2G 087018-5G
102852-004EA 490.77 USD
102852-004 102852-006 102852-008
5-Oxo-5-(2-pyridyl)valeric acid ≥97%
5-Oxo-5-(2-pyridyl)valeric acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD01320176
CAS Number: 13672-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR