7-Oxononanoic acid ≥97%

Supplier: Matrix Scientific

087004-5G 087004-2G 087004-1G
102851-920EA 886.15 USD
102851-920 102851-924 102851-922
7-Oxononanoic acid ≥97%
7-Oxononanoic acid

Hazard: Irritant

Formula: C₉H₁₆O₃
MW: 172.23 g/mol
Melting Pt: 42…44 °C
MDL Number: MFCD01320161
CAS Number: 20356-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR