5-Cyclopropyl-5-oxovaleric acid ≥97%
Supplier: Matrix Scientific
086998-2G
086998-1G
086998-5G
102851-886EA
944.62
USD
102851-886
102851-884
102851-888
5-Cyclopropyl-5-oxovaleric acid ≥97%
5-Cyclopropyl-5-oxovaleric acid
Hazard: Irritant
Formula:
C₈H₁₂O₃ |
MDL Number:
MFCD01320152 CAS Number: 898766-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...