4-Cyclopentyl-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

086994-1G 086994-2G 086994-5G
102851-860EA 592.31 USD
102851-860 102851-864 102851-862
4-Cyclopentyl-4-oxobutyric acid ≥97%
4-Cyclopentyl-4-oxobutyric acid

Hazard: Irritant

Formula: C₉H₁₄O₃
MDL Number: MFCD01320143
CAS Number: 3400-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR