5-Cyclohexyl-5-oxovaleric acid ≥97%
Supplier: Matrix Scientific
086993-2G
086993-5G
086993-1G
102851-854EA
592.31
USD
102851-854
102851-858
102851-856
5-Cyclohexyl-5-oxovaleric acid ≥97%
5-Cyclohexyl-5-oxovaleric acid
Hazard: Irritant
Formula:
C₁₁H₁₈O₃ |
MDL Number:
MFCD01320140 CAS Number: 15971-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...