5-(2-Methoxyphenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

086982-2G 086982-1G 086982-5G
102851-790EA 944.62 USD
102851-790 102851-792 102851-788
5-(2-Methoxyphenyl)-5-oxovaleric acid ≥97%
5-(2-Methoxyphenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MDL Number: MFCD01311634
CAS Number: 105253-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR