4-(3-Bromophenyl)-4-oxobutyric acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(3-Bromophenyl)-4-oxobutanoic acid
086964-2G
086964-1G
086964-5G
102851-680EA
735.38
USD
102851-680
102851-684
102851-682
4-(3-Bromophenyl)-4-oxobutyric acid ≥97%
4-(3-Bromophenyl)-4-oxobutyric acid
Hazard: Irritant
Formula:
C₁₀H₉BrO₃ MW: 257.09 g/mol |
MDL Number:
MFCD00672056 CAS Number: 62903-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...