5-(4-Bromophenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

086963-2G 086963-5G 086963-1G
102851-674EA 306.15 USD
102851-674 102851-678 102851-676
5-(4-Bromophenyl)-5-oxovaleric acid ≥97%
5-(4-Bromophenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₁H₁₁BrO₃
Melting Pt: 128…130 °C
MDL Number: MFCD00672057
CAS Number: 35333-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR