3-(3-Methylphenyl)-1,1,1-trifluoro-2-propanone ≥97%
Supplier: Matrix Scientific
086953-2G
086953-1G
086953-5G
102851-618EA
706.15
USD
102851-618
102851-616
102851-614
3-(3-Methylphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(3-Methylphenyl)-1,1,1-trifluoro-2-propanone
Hazard: Irritant
Formula:
C₁₀H₉F₃O |
MDL Number:
MFCD03844194 CAS Number: 898787-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...