3-(4-Biphenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086938-2G 086938-1G 086938-5G
102851-530EA 520 USD
102851-530 102851-534 102851-532
3-(4-Biphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(4-Biphenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₁₅H₁₁F₃O
MDL Number: MFCD02260854
CAS Number: 898787-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR