1-Naphthyltrifluoromethyl ketone ≥97%
Supplier: Matrix Scientific
086918-2G
086918-1G
086918-5G
102851-410EA
520
USD
102851-410
102851-414
102851-412
1-Naphthyltrifluoromethyl ketone ≥97%
1-Naphthyltrifluoromethyl ketone
Hazard: Irritant
Formula:
C₁₂H₇F₃O Melting Pt: 25 °C |
MDL Number:
MFCD01320007 CAS Number: 6500-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...