3',4'-Dichloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3,4-Dichlorophenyl)-2,2,2-trifluoroethanone

086911-5G 086911-2G 086911-1G
102851-372EA 1521.54 USD
102851-372 102851-370 102851-368
3',4'-Dichloro-2,2,2-trifluoroacetophenone ≥97%
3',4'-Dichloro-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
MDL Number: MFCD01319995
CAS Number: 125733-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR