2,2,2,3',4',5'-Hexafluoroacetophenone ≥97%

Supplier: Matrix Scientific

086910-5G
102851-366EA 1560 USD
102851-366
2,2,2,3',4',5'-Hexafluoroacetophenone ≥97%
2,2,2,3',4',5'-Hexafluoroacetophenone

Hazard: Irritant

Formula: C₈H₂F₆O
MDL Number: MFCD01319994
CAS Number: 886369-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR