3',5'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(3,5-Dimethylphenyl)-2,2,2-trifluoroethanone
086908-1G
086908-2G
086908-5G
102851-350EA
347.69
USD
102851-350
102851-354
102851-352
3',5'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
3',5'-Dimethyl-2,2,2-trifluoroacetophenone
Hazard: Irritant/Refrigerate
Formula:
C₁₀H₉F₃O |
MDL Number:
MFCD01319988 CAS Number: 132719-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...