4'-(Methylthio)-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2,2-Trifluoro-1-(4-(methylthio)phenyl)ethanone
086902-1G
086902-5G
086902-2G
102851-314EA
347.69
USD
102851-314
102851-318
102851-316
4'-(Methylthio)-2,2,2-trifluoroacetophenone ≥97%
4'-(Methylthio)-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₉H₇F₃OS MW: 220.21 g/mol |
MDL Number:
MFCD01319976 CAS Number: 122243-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...