4'-Cyano-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(2,2,2-Trifluoroacetyl)benzonitrile
086894-5G
086894-2G
086894-1G
102851-266EA
552.31
USD
102851-266
102851-268
102851-270
4'-Cyano-2,2,2-trifluoroacetophenone ≥97%
4'-Cyano-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₉H₄F₃NO MW: 199.13 g/mol Melting Pt: 52…54 °C |
MDL Number:
MFCD01319971 CAS Number: 23516-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...