2'-Cyano-2,2,2-trifluoroacetophenone monohydrate ≥97%
Supplier: Matrix Scientific
086892-1G
086892-5G
086892-2G
102851-256EA
789.23
USD
102851-256
102851-254
102851-258
2'-Cyano-2,2,2-trifluoroacetophenone monohydrate ≥97%
2'-Cyano-2,2,2-trifluoroacetophenone monohydrate
Hazard: Irritant
Formula:
C₉H₆F₃NO₂ |
MDL Number:
MFCD08741512 CAS Number: 898787-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...