2-Phenylbenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

086885-2G 086885-1G 086885-5G
102851-212EA 387.69 USD
102851-212 102851-216 102851-214
2-Phenylbenzoylacetonitrile ≥97%
2-Phenylbenzoylacetonitrile

Hazard: Irritant

Formula: C₁₅H₁₁NO
MDL Number: MFCD02260809
CAS Number: 270084-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR