4'-Acetoxy-5-chlorovalerophenone ≥97%
Supplier: Matrix Scientific
086876-2G
086876-5G
086876-1G
102851-162EA
904.62
USD
102851-162
102851-158
102851-160
4'-Acetoxy-5-chlorovalerophenone ≥97%
4'-Acetoxy-5-chlorovalerophenone
Hazard: Irritant
Formula:
C₁₃H₁₅ClO₃ |
MDL Number:
MFCD07700127 CAS Number: 898786-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...