2-Acetylbiphenyl ≥97%

Supplier: Matrix Scientific

Synonyms: 1-([1,1'-Biphenyl]-2-yl)ethanone

086872-5G 086872-2G 086872-1G
102851-134EA 1223.08 USD
102851-134 102851-138 102851-136
2-Acetylbiphenyl ≥97%
2-Acetylbiphenyl

Hazard: Irritant

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Melting Pt: 54…57 °C
MDL Number: MFCD00053707
CAS Number: 2142-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR