4-(4-Methoxyphenyl)-4-oxobutyronitrile ≥97%

Supplier: Matrix Scientific

086870-2G 086870-1G 086870-5G
102851-124EA 560 USD
102851-124 102851-122 102851-126
4-(4-Methoxyphenyl)-4-oxobutyronitrile ≥97%
4-(4-Methoxyphenyl)-4-oxobutyronitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MW: 189.22 g/mol
MDL Number: MFCD00158889
CAS Number: 55234-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR