Cyclopentyl-2-(4-methylphenyl)ethyl ketone ≥97%
Supplier: Matrix Scientific
086854-2G
086854-1G
086854-5G
102851-030EA
2172.31
USD
102851-030
102851-028
102851-026
Cyclopentyl-2-(4-methylphenyl)ethyl ketone ≥97%
Cyclopentyl-2-(4-methylphenyl)ethyl ketone
Hazard: Irritant
Formula:
C₁₅H₂₀O |
MDL Number:
MFCD03843042 CAS Number: 898769-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...