3,4-Dichloro-2'-(4-methylpiperazinomethyl)benzophenone ≥97%

Supplier: Matrix Scientific

086839-1G 086839-5G 086839-2G
102850-938EA 1116.92 USD
102850-938 102850-936 102850-940
3,4-Dichloro-2'-(4-methylpiperazinomethyl)benzophenone ≥97%
3,4-Dichloro-2'-(4-methylpiperazinomethyl)benzophenone

Hazard: Irritant

Formula: C₁₉H₂₀Cl₂N₂O
MW: 363.29 g/mol
MDL Number: MFCD03842842
CAS Number: 898762-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR