Cyclopentyl-3-methylphenyl ketone ≥97%

Supplier: Matrix Scientific

086807-5G 086807-2G 086807-1G
102850-746EA 1116.92 USD
102850-746 102850-748 102850-744
Cyclopentyl-3-methylphenyl ketone ≥97%
Cyclopentyl-3-methylphenyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03841292
CAS Number: 85359-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR